HYDRANAL™ - NEXTGEN Coulomat A-FA, Anolyte for Coulometric Karl Fischer Titration, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34471-500ML 34471-6X500ML
HONC34471-500MLEA 442 EUR
HONC34471-500ML HONC34471-6X500ML
HYDRANAL™ - NEXTGEN Coulomat A-FA, Anolyte for Coulometric Karl Fischer Titration, Fluka™
Karl Fischer titranten en reagentia
Anolyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, preferred for cells with diaphragm, acetonitrile-based, free of alcohols (free of CMR substances)
  • Assay of water Max. 50 ppm
  • Number of samples Min. 2
Order Now
Specificaties
Environmentally Preferable