High recovery glass snap top microvials for <2 ml samples, level 3 high performance applications, SureSTART™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6PRV11-15V 6PRV11-15VT 6PRV11-S1V
HYPE6PRV11-15VEA 126 EUR
HYPE6PRV11-15V HYPE6PRV11-15VT HYPE6PRV11-S1V
High recovery glass snap top microvials for <2 ml samples, level 3 high performance applications, SureSTART™
flesjes
Choose Thermo Scientific™ SureSTART™ high recovery glass snap top microvials, performance level 3, to maximise injection volume when analysing <2 ml samples.
With outer dimensions of 12×32 mm, these clear and amber glass microvials have an inner V-shaped bottom to enable a relatively high total usable volume and a residual volume of 4 µl. This is a performance level 3 vial, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses.