SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6PME03C1SP 6PME03C1SPF 6PME03C1SPT 6PME03C1SSP 6PME03C1SSPF 6PME03C1SSPT 6PME04C1 6PME04C1F 6PME04C1T 6PME05F1 6PME04F1 6PME04F1BR
HYPE6PME03C1SPEA 123 EUR
HYPE6PME03C1SP HYPE6PME03C1SPF HYPE6PME03C1SPT HYPE6PME03C1SSP HYPE6PME03C1SSPF HYPE6PME03C1SSPT HYPE6PME04C1 HYPE6PME04C1F HYPE6PME04C1T HYPE6PME05F1 HYPE6PME04F1 HYPE6PME04F1BR
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications
flesjes
Use Thermo Scientific™ SureSTART™ performance level 3 inserts with 2 ml vials when analysing <2 ml samples to maximise the injection volume. These glass inserts have a small total volume capacity of <400 μl, with a minimum residual volume of 2 µl.
When placed and sealed inside a 2 ml vial, microsamples can be used in conventional autosampler trays. Choose from polyspring conical, pulled point conical, or flat bottom insert formats. Conical inserts are most effective with residual volumes of 2 to 4 μl. These inserts are performance level 3, which means they are designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses. They are specification certified to ensure physical dimensions and meet all requirements of US, EU, JPN Pharmacopeia.