MSCERT Screw vial and cap kits, level 3 High performance applications, SureSTART™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6PMSCK534W 6PMSCK535W 6PMSCK540W 6PMSCK541W 6PMSCK601W 6PMSCK611W 6PMSCK641W 6PMSCK651W
HYPE6PMSCK534WEA 112 EUR
HYPE6PMSCK534W HYPE6PMSCK535W HYPE6PMSCK540W HYPE6PMSCK541W HYPE6PMSCK601W HYPE6PMSCK611W HYPE6PMSCK641W HYPE6PMSCK651W
MSCERT Screw vial and cap kits, level 3 High performance applications, SureSTART™
flesjes
Meet your certification and documentation needs with our level 3 Thermo Scientific™ SureSTART™ MSCERT screw vial and cap kits. Select level 3 products for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity.
Choose from a variety of kits containing short thread screw vials with marking spots in clear or amber glass and polypropylene caps with silicone/PTFE septa designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. Select kits with SureStop™ vials and/or AVCS™ screw caps forMore Product Information