Glass screw top headspace vials, 10 ml, level 2 high-throughput applications, SureSTART™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6ASV10-1 6ASV10-1F 6ASV10-1T 6ASV10-2 6ASV10-2T
HYPE6ASV10-1EA 77.7 EUR
HYPE6ASV10-1 HYPE6ASV10-1F HYPE6ASV10-1T HYPE6ASV10-2 HYPE6ASV10-2T
Glass screw top headspace vials, 10 ml, level 2 high-throughput applications, SureSTART™
flesjes
Use Thermo Scientific™ SureSTART™ 10 ml glass screw top headspace vials, performance level 2, in your high-throughput volatile gas analyses. Our screw top headspace vials are a novelty in a crimp-neck dominated field.
They provide a ready-to-use, convenient solution that does not require crimping or decrimping tools. These are performance level 2 vials, ensuring high quality data with an uninterrupted workflow in high-throughput applications using GC and single or triple quadrupole MS systems. The headspace vials are made to withstand higher temperatures and internal pressures required for volatile gas analyses. They have very low metal content, extractables, and leachables and conform to high standards to meet your regulatory requirements.