Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6ASV20-1 6ASV20-1F 6ASV20-1T 6ASV20-2 6ASV20-2T
HYPE6ASV20-1EA 65.8 EUR
HYPE6ASV20-1 HYPE6ASV20-1F HYPE6ASV20-1T HYPE6ASV20-2 HYPE6ASV20-2T
Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
flesjes
Use Thermo Scientific™ SureSTART™ 20 ml glass screw top headspace vials in your high-throughput volatile gas analyses. Performance level 2 vials ensure high quality data with an uninterrupted workflow in high-throughput applications using GC and single or triple quadrupole MS systems.
These headspace vials are made to withstand higher temperatures and internal pressures required for volatile gas analyses. They have very low metal content, extractables, and leachables and conform to high standards to meet your regulatory requirements. These vials are specification certified to ensure their physical dimensions and autosampler compatibility.