Specification certified screw vial and cap kits, level 2 high-throughput applications, SureSTART™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6AK592W 6AK594W 6AK599W 6AK78BW 6AK80W 6AK82W 6AK86W 6AK92W 6AK93BW 6AK94W 6AK95W 6AK186W 6AK595W 6AK195W 6AK192W 6AK180W 6AK188W 6AK196W
HYPE6AK592WEA 59.2 EUR
HYPE6AK592W HYPE6AK594W HYPE6AK599W HYPE6AK78BW HYPE6AK80W HYPE6AK82W HYPE6AK86W HYPE6AK92W HYPE6AK93BW HYPE6AK94W HYPE6AK95W HYPE6AK186W HYPE6AK595W HYPE6AK195W HYPE6AK192W HYPE6AK180W HYPE6AK188W HYPE6AK196W
Specification certified screw vial and cap kits, level 2 high-throughput applications, SureSTART™
flesjes
Meet your certification and documentation needs with our level 2 Thermo Scientific™ SureSTART™ specification certified screw vial and cap kits. Select level 2 products for high throughput applications, when robustness and reproducibility are key.
Choose from a variety of kits containing short thread screw vials with marking spots in clear or amber glass and polypropylene caps with silicone/PTFE septa of various hardness values designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. Select kits with SureStop™ vials and/or AVCS™ screw caps for effective seals, consistent data, and increased sample throughput.