Glass snap top vials, 2 ml, level 2 high-throughput applications, SureSTART™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6ARV11-1P 6ARV11-1PF 6ARV11-1PT 6ARV11-2P 6ARV11-2PF 6ARV11-2PT
HYPE6ARV11-1PEA 18.4 EUR
HYPE6ARV11-1P HYPE6ARV11-1PF HYPE6ARV11-1PT HYPE6ARV11-2P HYPE6ARV11-2PF HYPE6ARV11-2PT
Glass snap top vials, 2 ml, level 2 high-throughput applications, SureSTART™
flesjes
Choose Thermo Scientific™ SureSTART™ 2 ml glass snap vials, performance level 2, to ensure high quality data with an uninterrupted workflow in high-throughput applications using GC, HPLC/UHPLC, and single or triple quadrupole MS systems.
Use level 2 vials for routine QA/QC applications, where the analyte levels are achievable or close to the method detection level. Available in clear and amber glass, snap top vials have very low metal content, extractables, and leachables and conform to high standards so that you can to meet your regulatory requirements. They are specification certified to ensure their physical dimensions and autosampler compatibility.