Glass crimp top vials, 2 ml, level 3 high performance applications, SureSTART™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6PCV11-1P 6PCV11-1PF 6PCV11-1PT
HYPE6PCV11-1PEA 21.8 EUR
HYPE6PCV11-1P HYPE6PCV11-1PF HYPE6PCV11-1PT
Glass crimp top vials, 2 ml, level 3 high performance applications, SureSTART™
flesjes
Choose Thermo Scientific™ SureSTART™ 2 ml glass crimp top vials, performance level 3, for high performance applications, when sensitivity is a must. Our level 3 vials are our highest performance vials, designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding GC-MS. These 2 ml clear glass crimp vials are specification certified to ensure their physical dimensions and autosampler compatibility. Use 11 mm crimp caps and septa to seal these vials. Seal and remove your crimp caps using Thermo Scientific manual and electronic crimpers andMore Product Information