High recovery glass screw top 1,7 ml microvials for <2 ml samples, level 3 high performance applications, SureSTART™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6PSV9-V1 6PSV9-V1F 6PSV9-V1T
HYPE6PSV9-V1EA 289 EUR
HYPE6PSV9-V1 HYPE6PSV9-V1F HYPE6PSV9-V1T
High recovery glass screw top 1,7 ml microvials for <2 ml samples, level 3 high performance applications, SureSTART™
flesjes
Choose Thermo Scientific™ SureSTART™ 1,7 ml high recovery glass screw top microvials, performance Level 3, when you need to maximise the injection volume for <2 ml samples.
With outer dimensions of 12×32 mm, these clear or amber glass microvials have an internal conical reservoir that provides a total usable volume of 1,5 ml and a residual volume of 4 µl. This is a Performance Level 3 vials, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses.