Caps for 2 ml glass crimp top vials, level 2 high-throughput applications, SureSTART™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6ACC11RT1 6ACC11RT1F 6ACC11RT1T 6ACC11ST1 6ACC11ST1F 6ACC11ST1T
548-2691EA 22.4 EUR
548-2691 548-2692 548-2693 548-2694 548-2695 548-2696
Caps for 2 ml glass crimp top vials, level 2 high-throughput applications, SureSTART™
Sluitingen Sluitingen voor chromatografievials
Choose Thermo Scientific™ SureSTART™ 2 ml glass crimp top vials, performance level 2, to ensure high quality data with an uninterrupted workflow in high-throughput applications using GC, HPLC/UHPLC, and single or triple quadrupole MS systems.
Use these vials for routine QA/QC applications, where the analyte levels are achievable or close to the method detection level. They have very low metal content, extractables, and leachables and conform to high standards so that you can meet your regulatory requirements. They are specification certified to ensure their physical dimensions and autosampler compatibility.