Caps for 2 ml glass crimp top vials, level 1 everyday analysis, SureSTART™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CHCC11-10 CHCC11-10F CHCC11-10T CHCC11-30 CHCC11-30F CHCC11-30T
HYPECHCC11-10EA 11.9 EUR
HYPECHCC11-10 HYPECHCC11-10F HYPECHCC11-10T HYPECHCC11-30 HYPECHCC11-30F HYPECHCC11-30T
Caps for 2 ml glass crimp top vials, level 1 everyday analysis, SureSTART™
Sluitingen Sluitingen voor chromatografievials
Use performance level 1 Thermo Scientific™ SureSTART™ 2 ml glass crimp top vials for everyday chromatography analysis. They provide an affordable choice and can be used with all GC and HPLC/UHPLC instrument types.
These 2 ml glass crimp top vials are specification certified to ensure autosampler compatible physical dimensions. Choose from clear and amber glass. The vials meet all requirements of US, EU, and JPN pharmacopia. Seal these crimp vials with 11 mm aluminium crimp caps and septa using a manual or electronic crimper tool.