Caps for 20 ml glass crimp top headspace vials, level 1 everyday analysis, SureSTART™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CHCC20-1 CHCC20-1F CHCC20-1M CHCC20-1MT CHCC20-1T CHCC20-2 CHCC20-2F CHCC20-2M CHCC20-2MT CHCC20-2T
HYPECHCC20-1EA 48.7 EUR
HYPECHCC20-1 HYPECHCC20-1F HYPECHCC20-1M HYPECHCC20-1MT HYPECHCC20-1T HYPECHCC20-2 HYPECHCC20-2F HYPECHCC20-2M HYPECHCC20-2MT HYPECHCC20-2T
Caps for 20 ml glass crimp top headspace vials, level 1 everyday analysis, SureSTART™
Sluitingen Sluitingen voor chromatografievials
Use performance level 1 Thermo Scientific™ SureSTART™ 20 ml glass crimp top vials for everyday chromatography analysis. They provide an affordable choice and can be used with all headspace GC instrument types.
These headspace vials are made of superior quality 1st hydrolytic class glass (Type 1) and have a thicker glass wall to withstand higher temperatures and internal pressures for the analysis of volatile gas in the vial’s headspace. The vials are specification certified to ensure autosampler compatible physical dimensions, and meet all requirements of US, EU, and JPN pharmacopia. SealMore Product Information