Crimp caps, 20 mm, level 2 high-throughput applications, SureSTART™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6ACC20-PFT 6ACC20-PFTT 6ACC20-ST1 6ACC20-ST1F 6ACC20-ST1T 6AMCC20-ST3 6AMCC20-ST3T 6AMCC20-STH 6AMCC20-STHF 6AMCC20-STHT 6APRCC20-ST1 6APRCC20-ST1F 6APRCC20-ST1T 6ACC20-SA3 6PMCC20-SA3
548-2697EA 119 EUR
548-2697 548-2699 548-2700 548-2701 548-2702 548-2715 548-2717 548-2718 548-2719 548-2720 548-2721 548-2722 548-2723 HYPE6ACC20-SA3 HYPE6PMCC20-SA3
Crimp caps, 20 mm, level 2 high-throughput applications, SureSTART™
Sluitingen Sluitingen voor chromatografievials
Use Thermo Scientific™ SureSTART™ 20 mm crimp caps with crimp vials that have a 20 mm opening, including our SureSTART 10 ml and 20 ml glass crimp top headspace vials (level 2). Select level 2 products for high-throughput applications, when robustness and reproducibility are key.
Choose from aluminium and magnetic steel caps with silicone/PTFE or butyl/PTFE septa of various hardness values designed to reduce autosampler needle issues. Caps with pressure release technology will split to allow the safe release of high internal vial pressures. Use Thermo Scientific™ manual or electronicMore Product Information