High-capacity toploading laboratory balances, Cubis® II MCA
Leverancier: Sartorius Balances
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MCA32202P2S000 MCA70201S2S000 MCA50201S2S000 MCA36201S2S000 MCA36201P2S000 MCA20201S2S000 MCA70200S2S000 MCA36200S2S000 MCA32202P2099 MCA70201S2099 MCA50201S2099 MCA36201S2099 MCA36201P2099 MCA20201S2099 MCA70200S2099 MCA36200S2099
SARBMCA32202P2S000EA 15350 EUR
SARBMCA32202P2S000 SARBMCA70201S2S000 SARBMCA50201S2S000 SARBMCA36201S2S000 SARBMCA36201P2S000 SARBMCA20201S2S000 SARBMCA70200S2S000 SARBMCA36200S2S000 SARBMCA32202P2099 SARBMCA70201S2099 SARBMCA50201S2099 SARBMCA36201S2099 SARBMCA36201P2099 SARBMCA20201S2099 SARBMCA70200S2099 SARBMCA36200S2099
High-capacity toploading laboratory balances, Cubis® II MCA
Balansen en Weegschalen
Cubis® II MCA high-capacity toploading balances with advanced user interface offer a maximum load between 11,2 and 70,2 kg and readability between 0,1 and 1 g provide the ideal model for every application.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status centre notifications
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Cubis® II balance series from Sartorius.
All weighing modules of the Cubis® II balance are designed for intuitive operation, further aided by intelligent diagnostic systems. These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement. It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
These MCA models with advanced user interface have a high resolution colour touch screen plus holders for ergonomic sample handling and automatic motorised levelling. The balances feature innovative draft shield technology, gesture sensor and climate monitoring. With complete traceability the balances help achieve 21 CFR Part 11 and EU Annex 11 compliance.
Bestelinformatie: There are different optional software packages, each contains applications or functions to enhance and customise the balance for the users’ workflows. The all inclusive QApps package (denoted by QP99) includes all 4 of these packages: Pharma; Advanced Applications; Utilities and Connectivity.