Analytical laboratory balances, Cubis® II MCE
Leverancier: Sartorius Balances
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MCE224S2S00-A MCE524S2S00-U MCE524P2S00-U MCE324S2S00-U MCE324P2S00-U MCE224S2S00-U MCE124S2S00-U MCE524S2S00-A MCE524P2S00-A MCE324S2S00-A MCE324P2S00-A MCE124S2S00-A MCE524S2S00-I MCE524P2S00-I MCE324S2S00-I MCE324P2S00-I MCE224S2S00-I MCE124S2S00-I MCE224S-U-MTS MCE225S-I-3MTS
SARBMCE224S2S00-AEA 12010 EUR
SARBMCE224S2S00-A SARBMCE524S2S00-U SARBMCE524P2S00-U SARBMCE324S2S00-U SARBMCE324P2S00-U SARBMCE224S2S00-U SARBMCE124S2S00-U SARBMCE524S2S00-A SARBMCE524P2S00-A SARBMCE324S2S00-A SARBMCE324P2S00-A SARBMCE124S2S00-A SARBMCE524S2S00-I SARBMCE524P2S00-I SARBMCE324S2S00-I SARBMCE324P2S00-I SARBMCE224S2S00-I SARBMCE124S2S00-I 611-5826 611-5832
Analytical laboratory balances, Cubis® II MCE
Balansen en Weegschalen
Cubis® II MCE analytical balances with essential user interface offer a maximum weighing capacity between 120 and 520 g in combination with readability of 0.1 mg.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status centre notifications
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Cubis® II balance series from Sartorius.
All weighing modules of the Cubis® II balance are designed for intuitive operation, further aided by intelligent diagnostic systems. These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement. It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
These MCE models with essential user interface have a black and white touch screen plus holders for ergonomic sample handling and automatic motorised levelling. The balances feature innovative draft shield technology, gesture sensor and climate monitoring. With complete traceability the balances help achieve 21 CFR Part 11 and EU Annex 11 compliance.
Choice of draft shield denoted by model name suffix: manual glass doors with large draft shield (U), motorised draft shield (A) or motorised with built-in de-ioniser (I) for effective removal of static charges.