Thermoplastic duct
Leverancier: LABCONCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7027200 5602000 4708600
LBCP7027200EA 0 EUR
LBCP7027200 LBCP5602000 LBCP4708600
Thermoplastic duct
Ducting Supplies
PVC exhaust duct is type one, unplasticised, schedule 40, lightweight, and corrosion-resistant.
A female duct coupling is required to join two sections. Connections can be made using solvent cement. PVC is rigid and may be cut without the use of special tools.