Cleanroom overalls, Tyvek® IsoClean®, Model IC183B TS
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Tyvek® IsoClean®
DUPOIC183BWHTS/XLEA 388 EUR
DUPOIC183BWHTS/XL DUPOIC183BWHTS/2XS DUPOIC183BWHTS/7XL DUPOIC183BWHTS/5XL DUPOIC183BWHTS/6XL DUPOIC183BWHTS/M DUPOIC183BWHTS/L DUPOIC183BWHTS/3XL DUPOIC183BWHTS/4XL DUPOIC183BWHTS/XS DUPOIC183BWHTS/2XL DUPOIC183BWHTS/S
Cleanroom overalls, Tyvek® IsoClean®, Model IC183B TS
Wegwerpkleding
These unhooded coveralls, model IC183BWH TS, are made of Tyvek®, a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability, and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
  • Unhooded overall with bound neck
  • Ultra-low linting fabric
  • Tyvek® covered elasticated thumb loops
  • Tunnelled elastic at wrists and ankles; stitched-in elastic waist
  • Front zipper closure with storm flap
  • Optimised shape and fit allowing full freedom of movement
  • Aseptically folded
  • Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
  • Suitable for use in GMP class A/B (ISO Class 5) cleanrooms
All DuPont™ Tyvek® IsoClean® sterile accessories (option code TS) are packed in a dual barrier packaging system, consisting of an inner and outer easy-tear cleanroom bag.
Garments made of Tyvek® IsoClean® are used in the biotech, pharmaceutical, medical device manufacturing, and electronics industries, as well as in other critical or controlled environments.
Certificaten: Type 5B/6B, EN 1073-2 (Class 2), EN 14126, Cat. III.
Verpakking: Individually double packed. Two polyethylene liners. Cardboard box. Packed in an ISO Class 5 certified cleanroom.