Cleanroom lab coats, Tyvek® IsoClean®, Model IC270B WS
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IC270BWHWS/S IC270BWHWS/M IC270BWHWS/L IC270BWHWS/XL IC270BWHWS/2XL IC270BWHWS/3XL IC270BWHWS/4XL
DUPOIC270BWHWS/SEA 393 EUR
DUPOIC270BWHWS/S DUPOIC270BWHWS/M DUPOIC270BWHWS/L DUPOIC270BWHWS/XL DUPOIC270BWHWS/2XL DUPOIC270BWHWS/3XL DUPOIC270BWHWS/4XL
Cleanroom lab coats, Tyvek® IsoClean®, Model IC270B WS
Wegwerpkleding
These lab coats with bound neck, model IC270BWH WS, deliver an ideal balance of protection, durability, and comfort. Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Elastic openings for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Adjustable snap at neck for better fit
  • Body-length jacket design with front snap closures for easy donning and doffing
  • Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
  • Suitable for use in GMP class A/B (ISO Class 5) cleanrooms
All DuPont™ Tyvek® IsoClean® sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy-tear cleanroom bag.
Garments and accessories made of sterile Tyvek® IsoClean® are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry, as well as in other critical or controlled environments.
Certificaten: PPE Category I.
Verpakking: Individually packed. Subgrouped by five in an outer bag. Two polyethylene liners. Cardboard box.