Cleanroom boot covers, Tyvek® IsoClean®, Model IC458B WS
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IC458BWHWS/M IC458BWHWS/S IC458BWHWS/L IC458BWHWS/XL
DUPOIC458BWHWS/MEA 333 EUR
DUPOIC458BWHWS/M DUPOIC458BWHWS/S DUPOIC458BWHWS/L DUPOIC458BWHWS/XL
Cleanroom boot covers, Tyvek® IsoClean®, Model IC458B WS
Wegwerpkleding
These overboots, model IC458BWH WS, are made of Tyvek®, which is composed of flash spun high density polyethylene, providing an ideal balance of protection, durability, and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
  • Ultra-low-linting fabric
  • Fixation ties
  • Ankle ties for secure fit
  • Gripper™ soles provide enhanced skid-resistance and durability
  • Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
  • Bound seams
  • Suitable for use in GMP class A/B (ISO Class 5) cleanrooms
Tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration.
All DuPont™ Tyvek® IsoClean® sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy-tear cleanroom bag.
Garments and accessories made of sterile Tyvek® IsoClean® are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry, as well as in other critical or controlled environments.
Certificaten: Partial body chemical protective clothing, Type PB [6-B], EN 14126, Cat. III.
Verpakking: 100 per box, individually packed in pairs. Subgrouped by 20 in an outer bag. Two polyethylene liners. Cardboard box.