Respirators, T200 Series
Leverancier: RPB SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17-200-22-CE 17-200-32-CE 04-852 17-722 17-732 17-712 17-811 17-815 17-817 15-860 17-513 04-831 04-841 16-526
RPBS17-200-22-CEEA 0 EUR
RPBS17-200-22-CE RPBS17-200-32-CE RPBS04-852 RPBS17-722 RPBS17-732 RPBS17-712 RPBS17-811 RPBS17-815 RPBS17-817 RPBS15-860 RPBS17-513 RPBS04-831 RPBS04-841 RPBS16-526
Respirators, T200 Series
Ademhalingsmaskers Ademhalingssystemen
The RPB T200 is a lightweight multi-purpose respirator designed specifically for the healthcare and life science industries, relieving workers of breathing difficulties and fatigue experienced with N95 use. Available with either a head harness or bump cap for head protection and multiple hood options, the T200 has the ability to adapt to different environments that need face protection and respiratory protection.
  • Customizable internal layout with four adjustable head harness points and moveable lens and air duct system
  • Deconable head suspension system and disposable hood
  • Soft inner neck seal for extra comfort
  • Moveable lens and airduct positioning to accommodate different facial features and optical aids
  • Breathing tube easily attaches to respirator and swivels to eliminate tangles; centralized location ensures even weight distribution
  • Operating temperature: −10 to +60 °C
  • Noise rating: NRR 21/SNR 28
TH3 respiratory protection. No face fit testing required.
Designed to allow the ears to be free for doctors to utilize stethoscopes, or converse easily with patients with full view of the wearers face to assist in patient relationship building and empathy. Large viewing window for exceptional downward and peripheral vision allowing for both verbal and non-verbal communication. The advanced cradle allows the visor distance to be adjusted away from the face for professions which required optical aids such as loops.
During use within contagious environments, the outer Tychem® 2000 hood can be easily decontaminated, or cost effectively replaced as required. Multiple hood versions are available, with varying degree of cape lengths, depending on application and environment. Customizable head suspension system includes an adjustable ratchet at the back and over-the-head straps to allow for a secure, individualized fit.
DuPont™ Tychem® 2000 available in face seal, medium length and full length hood options. Full length hood also available in DuPont™ Tychem® 4000.
Certificaten: EN 12941:1998+A2:2008. TH3P R SL. EN 166:2001. Class 1F3.
Informatie over toebehoren: Replacement Tychem 2000 Hood with built-in polycarbonate visor available for RPB T-Link and T200 Respirator head tops. Each hood has an additional colour, which allows the inner to be tucked into the wearers scrubs (or paint suit) to direct the expelled air into their suit for cooling airflow and/or to protect the cotton inner seal from external contamination. Use RPB 17-815 tear-off lenses to ensure perfect vision during shift, and to prolong the life of the hood. Tychem 2000 breathing tube cover for use with the RPB medical and pharmaceutical range of respirator head top breathing tubes. For use in environments where cleanability, decontamination, and infection control are of utmost importance. Helps keep the ribbed breathing tubes clean by allowing them to be easily wiped down and disinfected between shifts, or disposed of and replaced if appropriate.
Bestelinformatie: Includes PX5 PAPR breathing tube, and head cradle. PAPR sold separately.