Weld grind respirators starter kit with PX5 PAPR, Z4
Leverancier: RPB SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
15-018-11-EUUK 16-922 15-878 16-872 16-873 15-711 16-530
RPBS15-018-11-EUUKEA 0 EUR
RPBS15-018-11-EUUK RPBS16-922 RPBS15-878 RPBS16-872 RPBS16-873 RPBS15-711 RPBS16-530
Weld grind respirators starter kit with PX5 PAPR, Z4
Ademhalingsmaskers Ademhalingssystemen
The RPB Z4 w/ PX5 PAPR starter kit provides everything needed to perform industrial welding and grinding tasks with the utmost in safety but also now with unmatched comfort and further reduced cost in use.
  • Flip up welding visor during grinding, while maintaining respiratory protection
  • Optimum vision with a clear replaceable grinding lens, ANSI Z87+ (EN 166) approved
  • Lightweight head top weighing just 805 g with ADF and cape
  • Aerodynamic shell design deflects fumes away
  • Large auto darkening welding lens with a 1/1/1 rating (variable lens shade (9-13))
  • Replaceable impact and ADF lenses for ease of maintenance (optional magnifying lens available)
  • FR rated head cover and cape
  • Operating temperature: −5 to +55 °C
  • Noise rating: NRR: 21/SNR: 28
The Z4 has been designed to retain all the features of other well established brands, while making incremental improvements where possible, based on direct feedback from the market, specifically around removing unwanted weight off the head, improving comfort, increased protection from sparks on the top of the head, lowering the cost in use and adding a range of optional accessories.
Comfortable multi-adjustment head suspension system is ergonomically balanced to reduce fatigue. Dual air distribution system delivers 75% of air supply to breathe zone via a multi-directional outlet. The remaining 25% is directed over the grinding visor to prevent fogging.
Certificaten: EN 12941:1998+A2:2008 TH3/2 P R SL, EN 166:2001 Class 1B, EN 379:2003+A1:2009, EN 175:1997, EN 812:2012 (optional).
Informatie over toebehoren: RPB Comms link in helmet communication kit, for RPB Z-Link, T-Link and Z4 respirator ranges. Allows adaptation to head top for in-helmet communication. Connects directly to current walkie-talkie radio. Note: radio adaptor plug sold separately. Please contact RPB to confirm which radio adaptor is required. RPB PX5 Back harnesses assembly is fire retardant. Additional or replacement standard battery for RPB PX5 PAPR is Lithium Ion.
Leveringsinformatie: Includes 1×RPB Z4 weld grind respirator with FR face seal, 1×1:1:1 auto darkening filter (shade 3, 5, 9-13), 1×PX5 PAPR breathing tube, 1×RPB slim line ear defenders, 1×PX5 PAPR, 1×FR rated belt, 1×particulate filter, 1×pack of 10 prefilters, 1×spark arrestor, 1×lithium ion rechargeable battery, 1×charging bank with EUUK plugs and 1×flow tester.