Climate control device and belt, C40
Leverancier: RPB SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
03-502 04-322-50
RPBS03-502EA 0 EUR
RPBS03-502 RPBS04-322-50
Climate control device and belt, C40
Ademhalingsmaskers Ademhalingssystemen
RPB C40 Climate device assembly and belt. For use with RPB's range of SAR respiratory head top, including Z-Link, T-Link, and Nova 3.
  • Easily control temperature with the slide of a lever
  • Durable FR or easy cleaning belt
  • Made from lightweight, robust material
  • Increase or decrease the airflow by turning the flow control dial
  • Choose from a rangeof plugs to suit specific applications
  • Air inlet swivels 360° to ensure the device sits comfortably on the hip
  • Operating temperature: −10 to 60 °C
  • Flow rate: 510 to 708 L/min
Allows the user to connect to their compressed breathing airline and control not only the amount of airflow to the respirator between 170 to 330 L/min, but also control the temperature of the incoming air flow depending on personal preference. By simply sliding the temperature lever, the user can cool the incoming air by up to 29 °C, or heat the air by up to 18 °C from outside ambient air temperature. This ensures the user is perfectly comfortable, no matter what their environment, increasing wellbeing, comfort and safety.
Certificaten: EN 14594:2005 3B/4B.