Water Baths, Stainless Steel, Hydro Series
Leverancier: Lauda
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LAUDL002902EA 1160 EUR
LAUDL002902 LAUDL002957 LAUDL002901 LAUDL002956 LAUDL002989 LAUDL002904 LAUDL002959 LAUDL002903 LAUDL002958 LAUDL002906 LAUDL002905 LAUDL002907 LAUDL002991 LAUDL002990 LAUDL002960 LAUDL002993 LAUDL002992 LAUDL002953 LAUDL002986 LAUDL002955 LAUDL002900 LAUDL002988 LAUDL002987 LAUDL002954
Water Baths, Stainless Steel, Hydro Series
Baden Waterbaden, ongeroerd
Simple to operate water baths. Working temperature range is 25 to 100 °C.
  • TFT display shows all process data clearly
  • Setting and displaying temperature values in increments of 0.1 °C
  • Setting the timer's time values in hours (h) and minutes (min)
  • Membrane keypad with concise symbols
  • Digital setpoint adjustment
  • Adjustable trigger points for over- and under-temperature alarms up to max. ±10 K
  • Temperature display can be switched from °C to °F
  • 1-point temperature controller
  • Optical and acoustic alarm for low-level and over/under temperature as well as for sensor break/short-circuit
  • Electronic monitoring of the temperature controller
Hydro baths feature three application-specific timer functions. The baths have a convenient drain valve. The bath interior, cover, sieve plate and heating element are made of stainless steel. The stainless steel lid is double-walled with inner curvature, it insulates and retains heat as well as preventing condensate from dripping back into the vessels.
Models H 8 A and H 16 A have an integrated circulation system to improve spatial temperature stability.
Leveringsinformatie: Supplied with lid and perforated tray as standard.