96-Well Plates, Barcoded, for Vanquish™ UHPLC Systems, WebSeal™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60180-P103B 60180-P105B 60180-P133B 60180-P135B 60180-P201B 60180-P206B 60180-P207B 60180-P211B 60180-P216B 60180-P217B
HYPE60180-P103BEA 51 EUR
HYPE60180-P103B HYPE60180-P105B HYPE60180-P133B HYPE60180-P135B HYPE60180-P201B HYPE60180-P206B HYPE60180-P207B HYPE60180-P211B HYPE60180-P216B HYPE60180-P217B
96-Well Plates, Barcoded, for Vanquish™ UHPLC Systems, WebSeal™
Microplaten Deepwell-microplaten
WebSeal™ PP well plates for Vanquish™ UHPLC systems, provides advanced sample security and sample integrity.
  • SBS format and barcode enables automatic inventory in the vanquish autosampler
  • Easy inventory management through barcode reading with empty shelf-level detection and well plate verification
Monitor and manage an inventory of up to 9 deep well plates or 20 shallow well plates (15 mm height) in the vanquish charger with an additional three positions in the Vanquish autosampler, for a maximum of 8832 samples.
The barcodes indicate a well plateMore Product Information