D-Mannose 6-phosphate sodium salt ≥95%
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M3065-100MG M3065-1GM
SPCMM3065-100MGEA 384 EUR
SPCMM3065-100MG SPCMM3065-1GM
D-Mannose 6-phosphate sodium salt ≥95%
D-Mannose 6-phosphate sodium salt
Storage temperature - REFRIGERATE: Keep tightly closed.
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00135859
CAS nummer: 70442-25-0
Order Now
Specification Test Results
Assay Approx. 95 %