Corning® Disposable Anti-static Spatulas
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3003 3004 3005 3006 3007 3012 3013
734-1521EA 277.49 EUR
734-1521 734-1522 734-1523 734-1524 734-1525 734-1529 734-1530
Corning® Disposable Anti-static Spatulas
Spatels
These spatulas are designed to save time and to provide contamination-free samples. They eliminate the recycling and resterilizing necessary with reusable spatulas.
  • Antistatic
  • Sterile
  • RNase-/DNase-free
  • Nonpyrogenic
Each spatula features two different tips to meet a variety of sampling requirements.
Bestelinformatie: Available in five different configurations. Microspatulas available in two configurations.
Verpakking: Individually packaged.