Corning® Gosselin™ Autoclave Bags
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SA03-01 SA09-01 SA14-01 SA21-01 SA33-01
129-0132EA 170 EUR
129-0132 129-0135 129-0138 129-0141 129-0144
Corning® Gosselin™ Autoclave Bags
Zakken Sterilisatiezakken
These autoclave bags are made of PP.
  • Every bag is 40 µm thick
  • Thermal resistance of 134 °C
  • Wide range of volumes available (3000 to 33000 ml)
  • Nonsterile
Bags must not be tightly sealed to withstand the temperature recommended for 20 minutes in autoclave.