PURELL® Advanced Hygienic Hand Rub Refills
Leverancier: GOJO Industries - Europe Ltd
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2156-08 5476-02-EEU
115-3457EA 190 EUR
115-3457 115-0553
PURELL® Advanced Hygienic Hand Rub Refills
Huidverzorging Huiddesinfectiemiddelen Vloeibare en schuimende huiddesinfectiemiddelen
A no-rinse hygienic hand rub that feels great to use and is clinically proven to maintain skin health.
  • Enriched with skin conditioners that help maintain skin moisture and improve overall skin feel
  • Kills 99,99% of most common germs that may be harmful
  • 70% alcohol formulation is suitable for frequent use by hospital staff
  • Removable pump for easier recycling
  • CLEAN SEALED™ refill helps prevent contamination
  • Halal certified
Recommended for use on hands that are lightly soiled, and in conjunction with a hand hygiene regime.
GOJO SMART-FLEX™ bottles areMore Product Information