GOJO® Freshberry Foam Hand Soap Refills
Leverancier: GOJO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GOJO®
115-3170EA 54.9 EUR
115-3170 115-1524 115-3131 GOJO8816-03-EEU00 115-1517
GOJO® Freshberry Foam Hand Soap Refills
Huidverzorging Huiddesinfectiemiddelen Vloeibare en schuimende huiddesinfectiemiddelen
Spa-inspired foam hand wash which provides a creamy and delicate lather.
  • Formulation contains a moisturising agent from a natural origin
  • Leaves the skin moisturised up to 4 h and feeling silky
  • Pleasant botanical fragrance leaves the skin delicately fragranced after use
  • Dermatologically tested
  • Refill bottles made with 100% recycled PET
  • Halal certified
Suitable for use within general washroom facilities. Creamy foam with a pleasant fragrance contains an emollient agent and plant-based glycerin, and is dermatologically tested.
Certificaten: Ecolabel certified.
Bestelinformatie: Some of these refills are part of the GOJO free dispenser program. To receive a FREE of CHARGE manual dispenser, you need to purchase at least one refill. The manual dispensers available for this offer are GOJO® ADX-12™, GOJO® ADX-7™, GOJO® PRO TDX™ and GOJO® FMX™. If you want a FREE of CHARGE touch-free dispenser, you need to purchase one case consisting of two refills to receive a GOJO® TFX™ white and two cases of two refills each to receive a GOJO® TFX™ metallic.