Culture inserts, BRAND Insert 2in1, for 24-well plates, sterile, BIO-CERT® cell culture quality
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
782712 782713 782714 782715
734-3675EA 350 EUR
734-3675 734-3676 734-3677 734-3678
Culture inserts, BRAND Insert 2in1, for 24-well plates, sterile, BIO-CERT® cell culture quality
Microplaten Insertplaten en inserts
The BRAND Insert 2in1 is a cell culture insert used for skin model production, 3D epithelium cultures, chemotaxis analyses, transmigration assays, transport and polarity studies, and more.
  • Inserts can be used in a hanging or standing position
  • For 24-, 12-, and 6-well plates or culture dishes
  • Achieve uniform and controlled cell growth thanks to surface treatment, without coatings
  • Suitable for 2D and 3D cell cultures
In many cell culture tasks, or when switching from a standing to a hanging position, you can continue to use your current cell culture plates, saving additional validation steps. Thanks to both supporting feet and hooks, you can position the BRAND Insert 2in1 either way. Via its protein-like structure, the cellGrade™ plus surface provides optimal conditions for growing primary keratinocytes and many other cell types. With the practical multipacks, the right number of inserts is always close at hand. They can be opened row by row, so that the remaining inserts stay sterile. Besides the practical benefits, this also reduces waste.
Certificaten: Sterility Assurance Level (SAL) of 10⁻⁶.
Verpakking: Packaged individually or in practical multi-packs.