Cleanroom overalls with attached hood, CleanMax®
Leverancier: LAKELAND CLEANMAX APPAREL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CTL428CM-2X CTL428CM-3X CTL428CM-4X CTL428CM-5X CTL428CM-LG CTL428CM-MD CTL428CM-XL CTL428CM-SM
114-9528EA 450 EUR
114-9528 114-9529 114-9530 114-9531 114-9532 114-9533 114-9534 CLLLCTL428CM-SM
Cleanroom overalls with attached hood, CleanMax®
Wegwerpkleding
Clean manufactured disposable coveralls are individually packaged and folded to promote easier gowning with less excursions.
  • Clean manufactured in silicone-free environment
  • Latex-free
  • Tunnelled elastic on wrists with thumb loops
  • Zipper closure
  • For critical environments
  • Individually bagged
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration,More Product Information