Corning® Cap, Phenolic, PTFE Liner
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9998-15 9998-13 9998-18 9998-24 9998-241
212-7528EA 280.68 EUR
212-7528 212-7527 212-7529 CORN9998-24 CORN9998-241
Corning® Cap, Phenolic, PTFE Liner
Sluitingen Reageerbuissluitingen
This cap is fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving.
  • Available in different sizes
Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface which provides a highly inert sealing face.