Corning® Disposable Phenolic Threaded Screw Cap with Rubber Liners, Corning
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
99999-13 99999-15 99999-18
734-4255EA 328.71 EUR
734-4255 734-4256 734-4257
Corning® Disposable Phenolic Threaded Screw Cap with Rubber Liners, Corning
Sluitingen Reageerbuissluitingen
These disposable Corning® caps are made from phenolic resin and are autoclavable at 121°C
  • Available in different sizes
They are provided with a white rubber liner.