Disposable aseptic transfer cap for disposable spinner flask, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3562 3564 3565 3567 3558 3545 3528
734-4098EA 435.17 EUR
734-4098 734-4100 734-4101 734-4102 CORN3558 CORN3545 CORN3528
Disposable aseptic transfer cap for disposable spinner flask, Corning®
Sluitingen Flesdoppen
These disposable Corning® spinner flask aseptic transfer caps will both reduce contamination and enhance the productivity of your cell culture operations when adding or removing medium and cells.
  • Cap is made of high density polyethylene; autoclaving is not recommended
  • The venting port ends in a 25 mm diameter vent filter with a 0,2 µm pore PVDF membrane and a barbed fitting
  • Chemically resistant, heat sealable flexible tubing, ending in a male medical plastic coupler (MPC) connector closed with a female sealing cap
Aseptic transfer caps are sterile (10⁻⁶ SAL), individually double bagged and fit the side arms on 1 L Corning® disposable spinner flasks (Cat. No. 89089-754). Cap is made of high density polyethylene; autoclaving is not recommended. The venting port ends in a 25 mm diameter vent filter with a 0,2 µm pore PVDF membrane and a barbed fitting. The liquid transfer port has an internal end with a rigid polypropylene dip tube angled to reach the bottom of the flask and on the external side a ¹/₈" ID (3,2 mm), ¹/₄" OD chemically resistant, heat sealable flexible tubing, ending in a male Medical Plastic Coupler (MPC) connector closed with a female sealing cap.
Verpakking: Aseptic transfer caps are sterile (10⁻⁶ SAL), individually double bagged.