Corning® DeckWorks Reload System Starter Kits, Corning
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4134 4130 4131 4133 4132
613-1525EA 79.23 EUR
613-1525 613-1521 613-1522 613-1524 613-1523
Corning® DeckWorks Reload System Starter Kits, Corning
Pipettips
Corning® DeckWorks™ pipette tip system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials.
  • Certified free of RNase, DNase, pyrogens and PCR inhibitors
Corning DeckWorks starter kits contain one DeckWorks reload system and empty hinged racks (10 empty racks accompany 10, 200 and 300 µl volume tips; eight empty racks acommpany 1000 µl volume tips). DeckWorks reload systems are convenient and economical - reload decks are packaged in trasparent trays for easy tip identification, require no transfer devices, save space and minimize packaging waste. Although the pipette tips are made from virgin medical-grade resins, the reload decks and rack bases are manufactured from 100% recycled polypropylene. Nonsterile decks and racks are steam autoclavable.
Both the hinged racks and relaod decks within this system are manufactured with recycled plastic. The entire DeckWorks system is comprised of 7 sizes of graduated pipette tips which are longer than standard tips to minimize the contamination issues often associated with traditional pipette tip styles. Sizes include 10, 20, 100, 200, 200, extended length, 300 and 1000 µl tips. The pipette tip s are made in a controlled molding environment to ensure accuracy and precision across all tip volumes. Medical grade resins and production excellence allow for full certification as: nonpyrogenic, human DNA-free, RNase-/DNase-free, sterility assuarance level 10⁻³. All tips are manufactured from polypropylene.
Bestelinformatie: Corning DeckWorks pipette tips are also available in bulk, hinged rack, barrier hinged rack, reload, and tip station configurations.