Cleanroom overalls, sterile, CleanMax®
Leverancier: LAKELAND CLEANMAX APPAREL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CTL417CS-2X CTL417CS-3X CTL417CS-4X CTL417CS-5X CTL417CS-LG CTL417CS-MD CTL417CS-XL
114-9591EA 493 EUR
114-9591 114-9592 114-9523 114-9524 114-9525 114-9526 114-9527
Cleanroom overalls, sterile, CleanMax®
Wegwerpkleding
CleanMax® Clean and Sterile Disposable Coveralls with Elastic Wrist/Ankle, Lakeland Industries
  • Clean and sterile
  • Manufactured in silicone-free environment
  • Latex-free
  • Tunnelled elastic on wrists (with thumb loops), ankles, and back half of waist
  • Individually double-bagged
  • Certificate of radiation included
  • Gamma radiation indicator dots on each package
CleanMax® cleanroom garments provide comfort, quality, and protection. CleanMax® sterilized garments are both clean manufactured and sterile (not all sterile garments are clean manufactured).
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunnelled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Certificaten: Compatible for ISO Class 4 – 8 cleanrooms and all controlled environments.