96-Well Polypropylene Storage Blocks, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3956 3957 3960 3961 3959 3958
734-1675EA 420.62 EUR
734-1675 734-1676 734-1679 734-1680 734-1678 734-1677
96-Well Polypropylene Storage Blocks, Corning®
Microplaten HTS-microplaten
Plates are available in V-bottom and round-bottom formats to meet a wide variety of assay and storage needs.
Robolid combines the sealing ability of a storage mat with the rigidity of a plastic lid and is designed for repeated application and removal by automation and to prevent short-term evaporation. Robolid also features silicone sealing plugs for organic solvent resistance and low extractables.
All sterile products are certified RNase- and DNase-free. Built to the industry standard footprint.
Bestelinformatie: Storage mats and sealing tapes sold separately.