INTERCEPT™ HyFlex® 11-251 Cut Resistant Sleeves with Thumbslot
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-251/18N 11-251/18W
ANSE11-251/18NEA 450 EUR
ANSE11-251/18N ANSE11-251/18W
INTERCEPT™ HyFlex® 11-251 Cut Resistant Sleeves with Thumbslot
Werkkleding
The most comfortable sleeve with INTERCEPT™ technology.
  • 11-251 sleeves with thumb slot come in different lengths and widths for optimal fit and comfort
  • Colour-coding will help with easy sorting after laundry
  • Sleeves are made of black yarn and are silicone-free
  • Special knitted bicep cuffs keep the sleeve in place
  • Compliant with FDA food handling regulations 21 CFR 177.1520 and 21 CFR 177.2600
This industrial sleeve with INTERCEPT™ cut resistance technology provides best-in-class cut protection and a soft and cool feel to the worker. The seamless design and the options of lengths and widths ensure an excellent fit and makes it an optimal choice for all day comfort. Sleeves are engineered to perform in combination with any HyFlex® cut resistant glove. Red and black over edge is designed to indicate wide (black) and narrow (red), for easy sorting and identification.