Industrial gloves, HyFlex® 11-422
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-422/10 11-422/7 11-422/8 11-422/9
ANSE11-422/10EA 169 EUR
ANSE11-422/10 ANSE11-422/7 ANSE11-422/8 ANSE11-422/9
Industrial gloves, HyFlex® 11-422
Handschoenen
Abrasion-resistant medium-duty work gloves, with fingerless glove tips for optimal dexterity. Ideal for the most delicate, intricate tasks.
  • PU/nitrile coating (made using a more eco-friendly water-based process) ensures EN level 4 abrasion resistance
  • Silicone-free design: risk of contaminant transfer to metal before painting is eliminated
  • Contact heat resistance up to 100 °C/212 °F
  • Seamless knitted liner (Fiber glass, Polyamide) enhances cut protection
This glove offers an indicator button (orange) which belongs to a range of colour codes to easily identify the glove cut level; 1 (green), 3 (orange), 5 (red).
Recommended for: Assembly of parts, electrical equipment operation and switching, press work.
Certificaten: EN 388:2016 (4X32B), Cat. II
Food contact approved