Cut-resistant industrial gloves with INTERCEPT™ technology, HyFlex® 11-550 series
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-550/10 11-550/11 11-550/6 11-550/7 11-550/8 11-550/9
ANSE11-550/10EA 142 EUR
ANSE11-550/10 ANSE11-550/11 ANSE11-550/6 ANSE11-550/7 ANSE11-550/8 ANSE11-550/9
Cut-resistant industrial gloves with INTERCEPT™ technology, HyFlex® 11-550 series
Handschoenen
Long lasting cut protection with INTERCEPT™ technology.
  • Liner provides excellent cut resistance and comfortable fit
  • Excellent protection from nuisance nicks and abrasion
  • Nitrile coating is engineered to provide workers with a secure grip in dry to light oil applications
  • Black palm coating for extended use life
Well-suited for a broad range of assembly applications and designed for dry or slightly oily applications where workers need cut protection. In some cases, protection from intermittent contact heat up to 212 °F (100 °C). Gloves are nitrile coated and cut-resistant, offering optimal comfort and a secure grip for enhanced productivity, safety and value.