Cut resistant gloves with INTERCEPT™ technology, HyFlex® 11-755
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-755/10 11-755/11 11-755/12 11-755/6 11-755/7 11-755/8 11-755/9
ANSE11-755/10EA 288 EUR
ANSE11-755/10 ANSE11-755/11 ANSE11-755/12 ANSE11-755/6 ANSE11-755/7 ANSE11-755/8 ANSE11-755/9
Cut resistant gloves with INTERCEPT™ technology, HyFlex® 11-755
Handschoenen
Ansell's lightest ANSI A5/ISO Level E cut resistant touchscreen glove.
  • Palm dipped PU coating, HPPE/Tungsten/Spandex/Polyester liner
  • Touchscreen compatible to ensure that workers do not need to change or remove gloves between tasks
  • Designed to fit the natural shape of the hand to help reduce hand fatigue
  • Clean and skin friendly, dermatologically approved (Dermatest®-approved)
Lightest weight ANSI A5/ISO Cut E glove featuring INTERCEPT™ Technology, these ultra-thin breathable gloves offer high cut resistance, while maintaining enhanced flexibility andMore Product Information
Order Now
Specificaties
Ambidexter/handspecifiek Handspecifiek
Environmentally Preferable
Voor eenmalig gebruik/herbruikbaar Herbruikbaar
Latex/latexvrij Latex
Afwerking Palm PU dipped
Steken per inch (Gauge) 18
Manchetstijl Gebreide manchet
EN-norm EN 388:2016 (3X41E)