HyFlex® 11-757 Cut-Resistant Industrial Gloves with INTERCEPT™ Technology
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-757/6 11-757/10 11-757/7 11-757/8 11-757/9
ANSE11-757/6EA 302 EUR
ANSE11-757/6 ANSE11-757/10 ANSE11-757/7 ANSE11-757/8 ANSE11-757/9
HyFlex® 11-757 Cut-Resistant Industrial Gloves with INTERCEPT™ Technology
Handschoenen
Ansell's lightest and thinnest ANSI A7/ISO level F cut resistant touchscreen glove.
  • Ultra-lightweight and ultra-thin breathable gloves are touchscreen compatible
  • High dexterity combined with unmatched comfort and precision while achieving excellent abrasion resistance
  • Tailored fit at the base of the pinky finger better matches natural hand contours
  • Dermatologically approved
Extreme cut protection featuring INTERCEPT™ Technology while maintaining enhanced flexibility and tactility. Ultra-thin ANSI cut level A7/ISO cut level F touchscreen compatible glove for enhanced productivity. Ultra-light design provides unmatched dexterity for handling and assembly of very sharp parts. A unique combination of ultra-lightweight comfort and extreme cut protection.