Industrial natural rubber latex gloves, AlphaTec® 87-029 series
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
87-029/8 87-029/9 87-029/7 87-029/10 87-029/11
ANSE87-029/8EA 111 EUR
ANSE87-029/8 ANSE87-029/9 ANSE87-029/7 ANSE87-029/10 ANSE87-029/11
Industrial natural rubber latex gloves, AlphaTec® 87-029 series
Handschoenen
Lightweight supported natural rubber gloves with knitted nylon liner.
  • Unique roughened finished with superb wet and dry grip performance
  • 13 gauge seamless nylon liner
  • High flexibility and excellent dexterity
  • Compliant with FDA and EU food handling requirements
  • AQL 1.5
Gloves offer good grip in both wet and dry environments due to the crinkle finish. They feature a polyamide seamless liner with wrinkle natural rubber latex coating on the hand, and smooth coating on the cuff.