Industrial gloves, HyFlex® 11-725
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-725/10 11-725/11 11-725/6 11-725/7 11-725/8 11-725/9
ANSE11-725/10EA 226 EUR
ANSE11-725/10 ANSE11-725/11 ANSE11-725/6 ANSE11-725/7 ANSE11-725/8 ANSE11-725/9
Industrial gloves, HyFlex® 11-725
Handschoenen
Comfortable, dexterous medium-duty work gloves, offering users moderate cut resistance.
  • INTERCEPT™ Cut Resistance Technology: Moderate cut resistance
  • HPPE/Lycra® liner: Heightened levels of comfort and flexibility, enabling workers to maintain productivity throughout the day
  • PU coating increases dexterity, while providing users a more secure grip in both dry and slightly oily environments
Recommended for: Assembling metal parts and components; assembly of white goods; metalwork, glass processing; handling sharp or irregular metal pieces; press injection molding.
Certificaten: EN 388:2016 (4342B)