Coveralls with hood, AlphaTec® 4000-GR
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GR40T-00111-08 GR40T-00111-09 GR40T-00301-07 GR40T-00301-06 GR40T-00301-02 GR40T-00301-03 GR40T-00301-04 GR40T-00301-05
ANSEGR40T-00111-08EA 446 EUR
ANSEGR40T-00111-08 ANSEGR40T-00111-09 ANSEGR40T-00301-07 ANSEGR40T-00301-06 ANSEGR40T-00301-02 ANSEGR40T-00301-03 ANSEGR40T-00301-04 ANSEGR40T-00301-05
Coveralls with hood, AlphaTec® 4000-GR
Werkkleding
Innovative multi-layer chemical barrier technology, offering Type 3/4/5 protection.
  • Exceptional protection; over 200 chemicals permeation tested, including chemical warfare agents
  • Designed to protect, typical innovative features include dual zip systems and double cuffs
  • Welded and taped seams, the highest barrier to liquids and particulates
  • Knitted inner cuffs and inner textile type fabric improves wearer comfort
A unique multi-layer barrier fabric renowned for lightweight, yet robust textile feel and exceptional barrier to organic and inorganic chemicals. Ultrasonically welded seams provide a strong liquid and particle barrier. Suits feature a two piece hood, elasticated hood, waist, double cuffs, and ankles, double zip closure, and double cuffs with knitted inner cuff.
Certificaten: Anti-static, tested according to EN 1149-5.