Industrial gloves, EDGE® 48-205 series
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
48-205/10 48-205/11 48-205/12 48-205/8 48-205/9
ANSE48-205/10EA 595 EUR
ANSE48-205/10 ANSE48-205/11 ANSE48-205/12 ANSE48-205/8 ANSE48-205/9
Industrial gloves, EDGE® 48-205 series
Handschoenen
An affordable solution for demanding extraction jobs.
  • High-performance fibres used in the construction of the palm protect hands from cut injuries
  • Back of hand TPR bumpers significantly reduce the risk of crush, pinch, and impact injuries
  • Excellent grip performance in dry, wet, and oily environments due to unique material on the palm
  • Reinforcement patches between thumb crotch and palm area improve longevity
  • Wide cuff with pull strap allows easy donning and doffing
Gloves offer superb cut resistant protection and are suitable for heavy-duty extractionMore Product Information