Industrial gloves, EDGE® 48-305
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
48-305/10 48-305/7 48-305/8 48-305/9
ANSE48-305/10EA 38.1 EUR
ANSE48-305/10 ANSE48-305/7 ANSE48-305/8 ANSE48-305/9
Industrial gloves, EDGE® 48-305
Handschoenen
EDGE® 48-305 industrial work gloves are the economical solution for medium weight applications requiring a good level of abrasion resistance along with a good grip in dry or wet environments.
  • Good dexterity and flexibility
  • Abrasion resistance is comparable to other latex coated gloves
  • Good level of tear resistance
  • Sure grip in dry and wet environments
  • Overedge colour code for easy size identification
EDGE® 48-305 is the economical choice for use in a wide range of handling and carrying applications, and are more durable than medium weight cotton or leather gloves.
Recommended for: General handling, packaging, shipping, street cleaning and refuse collection.
Certificaten: EN 388:2016 (2142B); Cat. II