Cut resistant industrial gloves, EDGE® 48-706
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
48-706/10 48-706/11 48-706/6 48-706/7 48-706/8 48-706/9
ANSE48-706/10EA 107 EUR
ANSE48-706/10 ANSE48-706/11 ANSE48-706/6 ANSE48-706/7 ANSE48-706/8 ANSE48-706/9
Cut resistant industrial gloves, EDGE® 48-706
Handschoenen
Thanks to their nitrile foam-coated palm, EDGE® 48-706 nitrile gloves offer a good grip and protection against cuts and abrasion in dry and slightly oily environments, making them ideal substitutes for heavy-duty leather and PU palm dipped, cut resistant knitted gloves.
  • Seamless liner (HPPE, Fiber glass, Nylon, Polyester, Spandex) for high wearing comfort without sacrificing dexterity
  • Nitrile foam coating for good grip in dry or slightly oily conditions
  • Made of FDA-compliant materials, these protective gloves are safe for food handling and packaging
Recommended for: General maintenance, hardware assembly, machining / stamping / cutting, molding, plastic injections, raw material handling, seat and frame build.
Certificaten: EN 388:2016 (4X43C); Cat. II