Oil resistant industrial gloves, EDGE® 48-929
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
48-929/10 48-929/11 48-929/6 48-929/7 48-929/8 48-929/9
ANSE48-929/10EA 125 EUR
ANSE48-929/10 ANSE48-929/11 ANSE48-929/6 ANSE48-929/7 ANSE48-929/8 ANSE48-929/9
Oil resistant industrial gloves, EDGE® 48-929
Handschoenen
Featuring a fully NBR-coated seamless liner with a rough palm, EDGE® 48-929 protective gloves offer a highly secure grip in oily environments, while protecting skin from injuries and oil contamination.
  • Medium-duty work gloves
  • Good combination of cut resistance, grip and oil-repellence
  • Excellent abrasion resistance
  • Double layer nitrile coating keeps hands dry and comfortable when handling oily parts and tools
  • Elastic knitwrist ensures a snug fit and optimal comfort
  • Gloves comply with FDA food handling requirements
Gloves are made of a seamless liner (Spandex/fiber glass/PE), fully dipped into NBR coating to protect skin from oil contamination. The rough NBR palm coating ensures a highly secure grip in oily environments. Designed for workers handling small and medium-sized sharp components coated with oil or lubricant.
Certificaten: EN 388:2016 (4X43C), Cat. II
Food contact approved.